Learn More
Aviva Systems Biology Corp LHX4 Antibody - middle region (ARP32424_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32424_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LHX4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LHX4. Alternate names: Gsh-4, Gsh4, CPHD4ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-54582