Learn More
Aviva Systems Biology Corp Lhx5 Antibody - middle region (ARP36908_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36908_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Lhx5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of mouse Lhx5. Alternate names: Lim2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-542