Learn More
Aviva Systems Biology Corp LHX6 Antibody - C-terminal region (ARP32553_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32553_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LHX6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human LHX6. Alternate names: LHX6.1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-54652