Learn More
Aviva Systems Biology Corp LHX8 Antibody - middle region (ARP47491_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47491_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LHX8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LHX8. Alternate names: Lhx7, LHX7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53517