Learn More
Aviva Systems Biology Corp Lhx8 Antibody - N-terminal region (ARP37081_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37081_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Lhx8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of mouse Lhx8. Alternate names: L3, Lhx7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-582