Learn More
Aviva Systems Biology Corp LHX9 Antibody - C-terminal region (ARP35966_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35966_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LHX9. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human LHX9. Alternate names: RP11-255O17.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53024