Learn More
Diasorin™ LIAISON NES™ FLU A/B, RSV, and COVID-19 Assay Kit, 10 Tests
Catalog No. 22011103 Shop All Diasorin Products
Change view
Click to view available options
Detectable Analytes:
Influenza A, Influenza B, COVID-19, RSV
Catalog No. Detectable Analytes
22-011-103 Influenza A, Influenza B, COVID-19, RSV
1 options
Catalog No. 22-011-103 Supplier Diasorin™ Supplier No. NES4450
 Restricted Item
Add to Cart
Add to Cart
The LIAISON NES™ FLU A/B, RSV, and COVID-19 Assay Kit is a rapid multiplex molecular test detecting Influenza A/B, RSV, and SARS-COV-2 in about 15 minutes. Fully automated on the compact LIAISON NES™ system, it enables simple setup, minimal hands-on time and reliable results during the patient visit.
  • Rapid molecular diagnostic test based on real-time PCR technology (RT-PCR)
  • Designed for the simultaneous and differential detection of Influenza A, Influenza B, Respiratory Syncytial Virus (RSV), and SARS-CoV-2
  • Ultra fast time to result of approximately 15 minutes.
  • The simple test setup and intuitive graphical user interface guide the user step-by-step, enabling confident operation even in decentralized care settings.
  • Minimized risk of errors thanks to streamlined workflow
  • Utilizes dry anterior nasal swabs, therefore avoiding the need for sample pipetting and its associated contamination risk
  • Requires non invasive sample type (anterior nasal swab)
  • Supports both professional collection and self-collection of the sample to be tested
  • Minimal hands-on time, thanks to the high automation on the LIAISON NES™ instrument.
  • The kit includes all components required to perform 10 individual patient tests, including anterior nasal swabs (10 units), reagent cartridges (10 units), and sample vials (10 units).
  • Each test is organized in individual, ready-to-use dividers, supporting efficient workflow and ease of use.
  • The kit is stable at room temperature, simplifying storage and handling requirements.
Specifications
CE Marker No
Clia Complexity Waived
Detectable Analytes Influenza A, Influenza B, COVID-19, RSV
Type Molecular Assay
Form Liquid, Lyophilized