Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diasorin™ LIAISON NES™ FLU A/B, RSV, and COVID-19 Control Swab Kit

Catalog No. 22011104 Shop All Diasorin Products
Change view
Click to view available options
Detectable Analytes:
Influenza A, Influenza B, COVID-19, RSV
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
22-011-104 Influenza A, Influenza B, COVID-19, RSV
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-011-104 Supplier Diasorin™ Supplier No. NES4460
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LIAISON NES™ Flu A/B, RSV, and COVID-19 Control Swab Kit containing color coded 10 positive control swabs (red) and 10 negative control swabs (green) to support quality control testing.

  • Workflow identical to a patient testing workflow, for consistency
  • Positive and negative controls are color coded to minimize risk of mistakes
  • Positive control consists in a swab coated with safe inactivated viral particles for Influenza A, Influenza B, Respiratory Syncytial Virus (RSV), and SARS-CoV-2
  • Breaking point to secure and contain the swab in the system during the reaction set up
  • The kit is stable at room temperature

Specifications

Detectable Analytes Influenza A, Influenza B, COVID-19, RSV
Sample Type Swab
Type Quality Control
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.