Learn More
Diasorin™ LIAISON NES™ Molecular Platform
Catalog No. 22011101 Shop All Diasorin Products
Change view
Click to view available options
Type:
Molecular Platform for Point-of-Care
Catalog No. Type
22-011-101 Molecular Platform for Point-of-Care
1 options
Catalog No. 22-011-101 Supplier Diasorin™ Supplier No. NES1001
 Restricted Item
Add to Cart
Add to Cart
LIAISON NES™ Instrument for molecular multiplex point-of-care testing of infectious diseases, highly automated, compact, portable and with advanced connectivity.
  • Compact and portable molecular platform for real-time PCR (RT-PCR) testing
  • Enables decentralized molecular testing
  • Small footprint
  • Highly automated and easy to use
  • Intuitive graphical user interface
  • Integrated barcode reader and internal barcode reader
  • Convenient status indicators (light and sound)
  • Advanced internet connectivity (Wi-Fi enabled, ethernet)
  • Lis and cloud connectivity
  • Minimal maintainance required, consisting of a simple cleaning procedure
Specifications
Type Molecular Platform for Point-of-Care
Sample Type Anterior Nasal Swab
Electrical Requirements 24 V
Dimensions (L x W x H) 10.2 x 4.3 x 9.5 in.