Learn More
DIAGNOSTICA STAGO INC Liatest D-Dimer
Catalog No. NC0233244
Encompass
The STA - Liatest D-Di kit is an immuno-turbidimetric assay for the quantitative determination of D-dimer in venous plasma (in 3.2 % sodium citrate) for use on STA-R, STA Compact and STA Satellite analyzers by professional laboratory personnel.
Catalog No. NC0233244 Supplier DIAGNOSTICA STAGO INC Supplier No. 00515
The STA - Liatest D-Di kit is an immuno-turbidimetric assay for the quantitative determination of D-dimer in venous plasma (in 3.2 % sodium citrate) for use on STA-R, STA Compact and STA Satellite analyzers by professional laboratory personnel.