Learn More
Sirius Automation Inc LibaryDoser Benchtop Lab Robot
SDP
Catalog No. 5000078210
Encompass
LibraryDoser automatically dispenses your powdered/granulated samples to individual target weight either one to many or many to many or multiplexed to a maximum of 24 individual sources and 96 target vials Information Weight data history ensures a forensic data trail and sample history for LIMS to maintain validation 0 1 or 0 01mg Weighing accuracy using Mettler Toledo Analytical Balance selections User interface is intuitive friendly can be file driven and is 21CFR11 capable This fast precise and proven robotic dispensing system fits in your standard hood most gloveboxes or our proprietary enclosure options Please ask for more information
Catalog No. 50-000-78210 Supplier Sirius Automation Inc Supplier No. LDSSTD02A
Add to Cart
Add to Cart
Request a Quote
LibraryDoser automatically dispenses your powdered/granulated samples to individual target weight either one to many or many to many or multiplexed to a maximum of 24 individual sources and 96 target vials Information Weight data history ensures a forensic data trail and sample history for LIMS to maintain validation 0 1 or 0 01mg Weighing accuracy using Mettler Toledo Analytical Balance selections User interface is intuitive friendly can be file driven and is 21CFR11 capable This fast precise and proven robotic dispensing system fits in your standard hood most gloveboxes or our proprietary enclosure options Please ask for more information