Learn More
Supply Solutions Liberase™ DH Research Grade, high Dispase concentration, 2x5mg
Catalog No. 501003341
Encompass
The most commonly used method for decellularization procedures, such as tissue dissociation and cell harvesting, is based on the use of proteolytic enzymes. Proteases disrupt the extracellular matrix of tissue to allow the release of individual cells or the harvesting of cultured cells for transfer. The goal of cell isolation is to maximize the yield of dissociated cells that are viable and functionally active.
Catalog No. 50-100-3341 Supplier Supply Solutions Supplier No. SIGMA 5401054001
The most commonly used method for decellularization procedures, such as tissue dissociation and cell harvesting, is based on the use of proteolytic enzymes. Proteases disrupt the extracellular matrix of tissue to allow the release of individual cells or the harvesting of cultured cells for transfer. The goal of cell isolation is to maximize the yield of dissociated cells that are viable and functionally active.