Learn More
Supply Solutions Liberase™ TH Research Grade, 2x5mg
Catalog No. 501005335
Encompass
Liberase TH Research Grade contains highly purified Collagenase I and Collagenase II. These two collagenase isoforms are blended in a precise ratio to each other and with a high concentration of Thermolysin, a non-clostridial neutral protease.
Catalog No. 50-100-5335 Supplier Supply Solutions Supplier No. SIGMA 5401135001
Liberase TH Research Grade contains highly purified Collagenase I and Collagenase II. These two collagenase isoforms are blended in a precise ratio to each other and with a high concentration of Thermolysin, a non-clostridial neutral protease.