Learn More
Supply Solutions Liberase™ TM Research Grade, medium Thermolysin concentration, 2x5mg
Catalog No. 501003280
Encompass
iberase TM Research Grade contains highly purified Collagenase I and Collagenase II. These two collagenase isoforms are blended in a precise ratio to each other, and with a medium concentration of Thermolysin, a non-clostridial neutral protease.
Catalog No. 50-100-3280 Supplier Supply Solutions Supplier No. SIGMA 5401119001
iberase TM Research Grade contains highly purified Collagenase I and Collagenase II. These two collagenase isoforms are blended in a precise ratio to each other, and with a medium concentration of Thermolysin, a non-clostridial neutral protease.