Learn More
Sigma Aldrich Fine Chemicals Biosciences Liberase TM Research Grade, 10 mg
Catalog No. NC1179175
Encompass
Liberase TM Research Grade contains highly purified Collagenase I and Collagenase II. These two collagenase isoforms are blended in a precise ratio to each other, and with a medium concentration of Thermolysin, a non-clostridial neutral protease.
Catalog No. NC1179175 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 5401119001
Liberase TM Research Grade contains highly purified Collagenase I and Collagenase II. These two collagenase isoforms are blended in a precise ratio to each other, and with a medium concentration of Thermolysin, a non-clostridial neutral protease.