Learn More
Elpro LIBERO CB Data Logger
SDP
Catalog No. NC1863810
Encompass_Preferred
This item must be ordered in multiples of 40 per supplier requirements. PDF Logger with internal NTC probe, single use, 2 temperature limits, alarm on MKT and logging time Connection: USB, generates PDF with alarm statistics and graph Expiry date according to label (auto-expiry)
Catalog No. NC1863810 Supplier Elpro Supplier No. 800002
This item must be ordered in multiples of 40 per supplier requirements. PDF Logger with internal NTC probe, single use, 2 temperature limits, alarm on MKT and logging time Connection: USB, generates PDF with alarm statistics and graph Expiry date according to label (auto-expiry)