Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Liberty Industrial Latex Gloves

Catalog No. 19121915D
Encompass
Encompass_Preferred
Change view
Click to view available options
Thickness (Metric):
0.12 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thickness (Metric)
19-121-915D 0.12 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-121-915D Supplier Liberty Supplier No. 2810WXL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Powder free latex gloves feature textured fingertips for increased dexterity.

  • Latex, industrial-grade
  • Powder free
  • Textured fingertips
  • Ambidextrous
  • 100 gloves per dispenser

Specifications

Chemotherapeutic Rated No
Applications Industrial
Color White
Disposable Single-use
Exterior Finish Partially Textured
Latex Free No
Material Latex
Powdered Powder Free
Size X-Large
Thickness (English) 5 mil
Thickness (Metric) 0.12 mm
Model 2810XL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.