Learn More
MSC Liberty Safety Cut Resistant Gloves, ANSI A2, 12 Pairs
Catalog No. 5000795257
Encompass
These Liberty Safety gloves are engineered for protection against cuts, punctures, and abrasions. They feature a durable engineered yarn construction with a microfoam nitrile coating on the palm and fingers, providing superior grip for both wet and dry applications.
  • Engineered yarn for high-visibility and seamless comfort
  • Microfoam nitrile coating on palm and fingers for superior grip
  • Seamless 18-gauge shell provides excellent dexterity, sensitivity, and tactility
  • Meets ANSI Cut Level A2, Abrasion 4, and Puncture 2 ratings
  • Durable construction suitable for multiple launderings
  • Colored cuffs for easy size identification
Catalog No. 50-007-95257 Supplier MSC Supplier No. 33646597
These Liberty Safety gloves are engineered for protection against cuts, punctures, and abrasions. They feature a durable engineered yarn construction with a microfoam nitrile coating on the palm and fingers, providing superior grip for both wet and dry applications.
  • Engineered yarn for high-visibility and seamless comfort
  • Microfoam nitrile coating on palm and fingers for superior grip
  • Seamless 18-gauge shell provides excellent dexterity, sensitivity, and tactility
  • Meets ANSI Cut Level A2, Abrasion 4, and Puncture 2 ratings
  • Durable construction suitable for multiple launderings
  • Colored cuffs for easy size identification