Learn More
MSC Liberty Safety Cut-Resistant Gloves, Cut A4, Size Large, 1 pair.
Catalog No. 5000687287
Encompass
Liberty Safety gloves feature a Hi-Vis Green seamless shell with a dark grey polyurethane palm coating, providing superior comfort and long-lasting protection. These gloves are highly resistant to abrasion and meet ANSI Cut Level A4 for high cut resistance.
  • Offers superior comfort and contains no fiberglass fibers
  • Features a high tensile strength to weight ratio
  • Highly resistant to abrasion, providing long-lasting protection
  • Provides non-slip grip, minimal lint, and is breathable
  • Durable, washable, and reusable
  • Colored hem cuffs allow for easy size identification
  • Made with HPPE primary material and a seamless lining
  • Polyurethane coating covers the palm and fingers
  • EN 388 rating of 4543 for performance
Catalog No. 50-006-87287 Supplier MSC Supplier No. 50712074
Liberty Safety gloves feature a Hi-Vis Green seamless shell with a dark grey polyurethane palm coating, providing superior comfort and long-lasting protection. These gloves are highly resistant to abrasion and meet ANSI Cut Level A4 for high cut resistance.
  • Offers superior comfort and contains no fiberglass fibers
  • Features a high tensile strength to weight ratio
  • Highly resistant to abrasion, providing long-lasting protection
  • Provides non-slip grip, minimal lint, and is breathable
  • Durable, washable, and reusable
  • Colored hem cuffs allow for easy size identification
  • Made with HPPE primary material and a seamless lining
  • Polyurethane coating covers the palm and fingers
  • EN 388 rating of 4543 for performance