Learn More
MSC Liberty Safety Gloves, Cut A4, Puncture 3, X-Large, 1 Pair
Catalog No. 5000768752
Encompass
These cut, puncture, and abrasion-resistant gloves feature a high-performance polyethylene fiber (HPPE) shell, a non-slip polyurethane coating on the palm and fingers, and a 13 gauge construction, providing reliable hand protection.
  • Cut, puncture, and abrasion-resistant
  • ANSI/ISEA Cut Resistance Level A4
  • ANSI/ISEA Puncture Resistance Level 3
  • ANSI/ISEA Abrasion Resistance Level 4
  • Constructed from high-performance polyethylene (HPPE)
  • Polyurethane coating on palm and fingers for non-slip grip
  • Features 13 gauge knit shell
  • High-visibility lime color with hi-vis green cuff
  • X-Large size
  • EN 388 rating: 4543
  • Elastic knit cuff
Catalog No. 50-007-68752 Supplier MSC Supplier No. 50712108
These cut, puncture, and abrasion-resistant gloves feature a high-performance polyethylene fiber (HPPE) shell, a non-slip polyurethane coating on the palm and fingers, and a 13 gauge construction, providing reliable hand protection.
  • Cut, puncture, and abrasion-resistant
  • ANSI/ISEA Cut Resistance Level A4
  • ANSI/ISEA Puncture Resistance Level 3
  • ANSI/ISEA Abrasion Resistance Level 4
  • Constructed from high-performance polyethylene (HPPE)
  • Polyurethane coating on palm and fingers for non-slip grip
  • Features 13 gauge knit shell
  • High-visibility lime color with hi-vis green cuff
  • X-Large size
  • EN 388 rating: 4543
  • Elastic knit cuff