Learn More
Medchemexpress LLC Libvatrep | 871814-52-7 | 99.5% | 305.33 | 50 MG
SDP
Catalog No. 5000435736
Encompass_Preferred
Vanilloid receptor antagonist 1 is a potent vanilloid receptor TRPV1 antagonist extracted from patent US8349852B2, compound B8.
  • For research use only.
  • Potent TRPV1 antagonist.
  • Not sold to patients.
Catalog No. 50-004-35736 Supplier Medchemexpress LLC Supplier No. HY11401750MG
Vanilloid receptor antagonist 1 is a potent vanilloid receptor TRPV1 antagonist extracted from patent US8349852B2, compound B8.
  • For research use only.
  • Potent TRPV1 antagonist.
  • Not sold to patients.