Learn More
Beckman Coulter Beckman Coulter PharmSpec software license for HIAC 9703+ (MPN 700060GP)
Catalog No. NC1904160
Encompass
Beckman Coulter PharmSpec software license (MPN 700060GP). Provides the PharmSpec data-acquisition and data-management application used with HIAC-series liquid particle counters to collect, analyze, and securely store particle-count data. Manufacturer materials reference PharmSpec 3; some reseller listings reference PharmSpec 3.5.
  • Provides data acquisition, analysis, and secure storage for liquid particle-counting results.
  • Supports 21 CFR Part 11 compliance and ALCOA data integrity principles.
  • Enables user authentication, access control, and encrypted data storage.
  • Includes installation media or package and system requirements documentation.
  • Allows scheduling of database backups to a secure network location.
Catalog No. NC1904160 Supplier Beckman Coulter Supplier No. 700060GP
Add to Cart
Add to Cart
Request a Quote
Beckman Coulter PharmSpec software license (MPN 700060GP). Provides the PharmSpec data-acquisition and data-management application used with HIAC-series liquid particle counters to collect, analyze, and securely store particle-count data. Manufacturer materials reference PharmSpec 3; some reseller listings reference PharmSpec 3.5.
  • Provides data acquisition, analysis, and secure storage for liquid particle-counting results.
  • Supports 21 CFR Part 11 compliance and ALCOA data integrity principles.
  • Enables user authentication, access control, and encrypted data storage.
  • Includes installation media or package and system requirements documentation.
  • Allows scheduling of database backups to a secure network location.