Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lichenan, ≥95%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN15523101 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15523101 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15523101 Supplier MP Biomedicals, Inc Supplier No. 0215523101

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1402-10-4
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.141
InChI Key CXKKSSKKIOZUNR-ZLUOBGJFSA-N
Synonym lichenin, 2s,3r,4r-2-hydroxymethyl-3,4-dihydro-2h-pyran-3,4,5-triol, licheningehalt
PubChem CID 439241
IUPAC Name (2S,3R,4R)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4,5-triol
SMILES C1=C(C(C(C(O1)CO)O)O)O

Specifications

Color Gray
Quantity 1 g
Formula Weight 488.442
Percent Purity ≥95%
Physical Form Powder
Chemical Name or Material Lichenan, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.