Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ 1,1,1,3,3,3-Hexafluoro-2-propanol, For GC derivatization, ≥99.8%, MilliporeSigma™ Supelco™

Catalog No. 111018566 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8566 10 mL
11-101-8364 10 x 1 mL
11-101-7984 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-8566 Supplier MilliporeSigma Supelco Supplier No. 5251710ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexafluoroisopropanol (HFP) is generally used to denature the native state of proteins, and it also stabilizes the a-helical conformation in unfolded peptides and proteins like ß-lactoglobulin and melittin.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00011651
Synonym HFP; Hexafluoroisopropanol

Specifications

Melting Point -4°C
Density 1.596 g/mL (at 25°C)
CAS 920-66-1
Boiling Point 59°C
Linear Formula (CF3)2 CHOH
Quantity 10 mL
Formula Weight 168.04
Percent Purity ≥99.8% (GC); ≥99.8%
Grade For GC derivatization
Product Line LiChropur
RTECSNumber UB6450000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.