Learn More
LiChropur™ 1-(Trimethylsilyl)imidazole, ≥94.0%, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Density | 0.956 g/mL (at 25°C) |
| CAS | 18156-74-6 |
| Boiling Point | 93°C to 94°C (14 mmHg) |
| Flash Point | 66°C |
| Packaging | Ampule |
| Linear Formula | C6H12N2 Si |
| Quantity | 10 x 1 mL |
| Formula Weight | 140.26 |
| Percent Purity | ≥94.0% |
| Grade | For GC derivatization |
| Show More |
Safety and Handling
| RTECSNumber | NI8700000 |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.