Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ 2,4-Dibromoacetophenone, 99.0%, MilliporeSigma™ Supelco™

Catalog No. 111019773 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9773 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9773 Supplier MilliporeSigma Supelco Supplier No. 680825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 99-73-0
Product Line LiChropur
Linear Formula BrC6H4 COCH2 Br
MDL Number MFCD00000200
Melting Point 108°C to 110°C; 108°C to 112°C
Synonym 4′-Bromophenacyl bromide
Type Organic Standard
Quantity 5 g
Formula Weight 277.94
Percent Purity ≥99% (HPLC)
Grade HPLC Derivatization
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.