Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ 2-Bromoacetophenone, For GC derivatization, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111019743 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9743 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9743 Supplier MilliporeSigma Supelco Supplier No. 7745010G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Bromoacetophenone is a commonly used derivatization agent for fatty acid detection in biological samples.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00000195
Synonym omega-Bromoacetophenone; Phenacyl bromide

Specifications

Melting Point 48°C to 51°C; 49°C to 50°C
CAS 70-11-1
Boiling Point 135°C (18 mmHg)
Flash Point 113°C - Closed Cup
Linear Formula C6H5 COCH2 Br
Quantity 10 g
Formula Weight 199.04
Percent Purity ≥99.0% (GC); ≥99.0%
Grade For GC derivatization
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.