Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ 4-(Dimethylamino)azobenzene-4-sulfonyl chloride, 98.0-102.0% (AT), MilliporeSigma™ Supelco™

Catalog No. 111018860 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8860 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8860 Supplier MilliporeSigma Supelco Supplier No. 89802500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00007444
Synonym 4-(4-Dimethylaminophenylazo)benzenesulfonyl chloride; DABS-Cl; Dabsyl chloride

Specifications

Melting Point 185°C
CAS 56512-49-3
Linear Formula (CH3)2 NC6H4 N=NC6H4 SO2 Cl
Quantity 500 mg
Formula Weight 323.80
Percent Purity 98.0 to 102.0% (AT)
Grade For HPLC derivatization
Product Line LiChropur
RTECSNumber DB8927800

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.