Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Acetic Anhydride, For GC derivatization, 99.0%, MilliporeSigma™ Supelco™

Catalog No. 111020385 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
10 x 1 mL
Packaging:
Glass Insert
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-102-0385 10 mL Glass Insert
11-101-9388 25 mL Glass Insert
11-101-9076 10 x 1 mL Glass Insert
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-102-0385 Supplier MilliporeSigma Supelco Supplier No. 9120410MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetic anhydride is an esterification agent which is corrosive to metals.

Reagent for acetic anhydride, acetyl, n-acetyl, n-acetyl-n, o-acetyl, acetate, aldononitrile acetate, aminoglycitol acetate, aryl acetate, permethyl acetyl, methyl ester, trifluoroacetyl, bis-trimethylsilyl, trimethylsilyl polyamino alcohol, and O-trimethylsilyl (TMS).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00008705
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
IUPAC Name acetyl acetate
SMILES CC(=O)OC(C)=O

Specifications

Melting Point -73°C
Density 1.08 g/mL
Boiling Point 138°C to 140°C
Packaging Glass Insert
Linear Formula (CH3CO)2 O
Refractive Index n20/D 1.390
Quantity 10 mL
Vapor Density 3.5 (vs. air)
Formula Weight 102.09
Vapor Pressure 4 mmHg (20°C)
Percent Purity ≥99.0% (GC)
Grade For GC derivatization
Physical Form Liquid
Show More Show Less
RTECSNumber AK1925000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.