Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Butylboronic Acid, For GC Derivatization, 96.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111014209 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-101-4209 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4209 Supplier MilliporeSigma Supelco Supplier No. 196675G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Derivatizing agent for gas chromatography

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4426-47-5
Molecular Formula C4H11BO2
Molecular Weight (g/mol) 101.94
MDL Number MFCD00002106
InChI Key QPKFVRWIISEVCW-UHFFFAOYSA-N
Synonym 1-Butaneboronic acid
IUPAC Name butylboronic acid
SMILES CCCCB(O)O

Specifications

Melting Point 90°C to 92°C; 90°C to 92°C
Packaging Glass Bottle
Linear Formula CH3(CH2)3 B(OH)2
Quantity 5 g
Formula Weight 101.94
Percent Purity ≥96.0% (T)
Grade For GC derivatization

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.