Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Dansyl chloride, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111020410 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
10 x 100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0410 100 mg
11-101-8299 10 x 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0410 Supplier MilliporeSigma Supelco Supplier No. 03641100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dansyl chloride is a fluorogenic reagent which is typically employed for the derivatization of amino acids.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00003985
Synonym 5-(Dimethylamino)naphthalene-1-sulfonyl chloride; DNSCl

Specifications

Melting Point 72°C to 74°C
CAS 605-65-2
Quantity 100 mg
Linear Formula C12H12 ClNO2 S
Solubility Information Acetone: 0.2 g/10 mL, clear to slightly turbid, yellow to very dark yellow-orange
Formula Weight 269.75
Percent Purity ≥99.0% (HPLC)
Grade HPLC
Product Line LiChropur
RTECSNumber QK3688000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.