Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Diethyl ethoxymethylenemalonate, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015718 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5718 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5718 Supplier MilliporeSigma Supelco Supplier No. 0568910ML

Chemical Identifiers

MDL Number MFCD00009148
Synonym DEEMM; Ethoxymethylenemalonic acid diethyl ester

Specifications

CAS 87-13-8
Density 1.07 g/mL (at 25°C)
Boiling Point 279°C to 281°C
Flash Point 144°C
Linear Formula C2H5 OCH=C(COOC2H5)2
Quantity 10 mL
Formula Weight 216.23
Percent Purity ≥98.0% (GC)
Product Line LiChropur
RTECSNumber OO1100000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.