Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Difluoroacetic Acid, MilliporeSigma™ Supelco™

Catalog No. 111022365 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
50 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2365 1 mL
11-102-2364 10 x 1 mL
11-102-2366 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-102-2365 Supplier MilliporeSigma Supelco Supplier No. 009221ML
Add to Cart
Add to Cart

Compared to trifluoroacetic acid (TFA), difluoroacetic acid (DFA) exhibits an improved sensitivity in mass spectral detection (ESI). It has a reduced effect on ion suppression in reversed-phase liquid chromatography (RPLC-MS), while keeping its strong chromatographic performance enhancing effects.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 381-73-7
Molecular Formula C2H2F2O2
Molecular Weight (g/mol) 96.03
MDL Number MFCD00004220
InChI Key PBWZKZYHONABLN-UHFFFAOYSA-N
Synonym DFA
IUPAC Name 2,2-difluoroacetic acid
SMILES OC(=O)C(F)F

Specifications

Melting Point -1°C (literature)
Density 1.526 g/mL (at 25°C (literature))
Boiling Point 132°C to 134°C (literature)
Flash Point 78°C (Closed Cup)
Linear Formula F2CHCO2H
Refractive Index n20/D 1.344 (literature)
Quantity 1 mL
Formula Weight 96.03
Percent Purity 97.5% to 102.5% (T)
Hazard Category H314
RTECSNumber AG9900000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.