Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Dihexylammonium acetate Solution, 0.5 M in Water, MilliporeSigma™ Supelco™

Catalog No. 111014047 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4047 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4047 Supplier MilliporeSigma Supelco Supplier No. 9364525ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ion-pairing agent suitable for LC/MS, HPLC, and ion chromatography analysis of nucleosides, nucleotides, and other compounds.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366793-17-1
Molecular Weight (g/mol) 245.40
MDL Number MFCD03093619
Synonym Dihexylamine acetate solution

Specifications

Density 0.992-0.996 g/mL (at 20°C)
Packaging Ampule
Linear Formula [(CH3(CH2)5]2 NH · CH3 COOH
Quantity 25 mL
Formula Weight 245.40
Grade For ion pair chromatography
Concentration or Composition (by Analyte or Components) 0.5 M aq. soln.
Physical Form Liquid
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.