Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Ethylenediaminetetraacetic acid disodium salt dihydrate, 99.0-101.0% (KT), MilliporeSigma™ Supelco™

Catalog No. 111015016 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5016 50 g
11-101-5015 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5016 Supplier MilliporeSigma Supelco Supplier No. 7988450G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

Synonym Disodium ethylenediaminetetraacetate dihydrate; EDTA disodium salt; EDTA-Na2; Edathamil; Edetate disodium salt dihydrate; Sequestrene Na2

Specifications

Melting Point 248°C
CAS 6381-92-6
Physical Form Crystalline Powder
Quantity 50 g
Linear Formula C10H14N2 Na2O8 · 2H2 O
Solubility Information Water: 0.1 M (at 20°C, clear, colorless)
Molecular Weight (g/mol) 372.24
Formula Weight 372.24
Percent Purity 99.0 to 101.0% (KT)
Grade HPLC
Product Line LiChropur
Chemical Name or Material Edetate Disodium Dihydrate
Show More Show Less
RTECSNumber AH4410000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.