Learn More
LiChropur™ Fmoc chloride, ≥99.0% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 28920-43-6 |
| Melting Point | 61°C to 64°C; 62°C to 64°C |
| Linear Formula | C15H11 ClO2 |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 258.70 |
| Formula Weight | 258.70 |
| Percent Purity | ≥99.0% (HPLC) |
| Grade | For HPLC derivatization |
| Physical Form | Solid |
| Product Line | LiChropur |
Safety and Handling
| RTECSNumber | LQ6250000 |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.