Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Heptafluorobutyric anhydride, For GC derivatization, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111018645 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8645 10 x 1 mL
11-101-8667 10 mL
11-101-8000 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-8645 Supplier MilliporeSigma Supelco Supplier No. 7725310X1ML
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00000432
Synonym HFAA; HFBA; Perfluorobutyric anhydride

Specifications

Melting Point -43°C
Density 1.674 g/mL at 20°C; 1.674 g/mL (at 20°C)
CAS 336-59-4
Boiling Point 108°C to 110°C
Packaging Ampule
Linear Formula (CF3 CF2 CF2 CO)2 O
Quantity 10 x 1 mL
Formula Weight 410.06
Percent Purity ≥99.0% (GC); ≥99.0%
Grade For GC derivatization
Product Line LiChropur
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.