Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Hydrindantin dihydrate for Stein-Moore-Chromatography, MilliporeSigma™ Supelco™

Catalog No. 111014803 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4803 10 g
11-101-4802 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4803 Supplier MilliporeSigma Supelco Supplier No. 5394010G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 5950-69-6
Product Line LiChropur
Linear Formula C18H14O8
MDL Number MFCD00149242
Melting Point 240°C
For Use With (Equipment) Stein-Moore-Chromatograph
Synonym 2,2′,3,3,3′,3′-Hexahydroxy-2,2′-biindan-1,1′-dione
Type Organic Standard
Quantity 10 g
Formula Weight 358.30

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.