Learn More
LiChropur™ Hydrogen Chloride - 1-butanol Solution, ∼3 M in 1-butanol, For GC Derivatization, MilliporeSigma™ Supelco™
Supplier:  MilliporeSigma Supelco 87472250MLF
 View more versions of this product
Catalog No. 11-101-8374
Add to Cart
Description
Description
Acylation, an alternative to silylation, is the conversion of compounds that contain active hydrogens (-OH, -SH and -NH) into esters, thioesters, and amides through the action of a carboxylic acid or derivative. The presence of a carbonyl group adjacent to the halogenated carbons enhances electron capture detector (ECD) response. Acylation has many benefits:. It improves the stability of compounds by protecting unstable groups.. It may confer volatility on substances such as carbohydrates or amino acids, which have so many polar groups that they are non-volatile and normally decompose on heating.. It assists in separations not possible with underivatized compounds.. Compounds are detectable at very low levels with an ECD.Hydrogen chloride-1-butanol is a reagent utilized to form fragmentation-directing derivatives for GC/MS analysis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7647-01-0
0.87 g/mL (at 20°C)
250 mL
MFCD00011324
GC Derivatization
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories
Promotions
Promotions
Product Title
Select an issue
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany