Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Hydrogen Chloride - 1-butanol Solution, ∼3 M in 1-butanol, For GC Derivatization, MilliporeSigma™ Supelco™

$119.00 - $453.00

Specifications

MDL Number MFCD00011324
Density 0.87 g/mL (at 20°C)
Grade GC Derivatization
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-9704
Accessories View Documents
MilliporeSigma Supelco
8747250MLF
50 mL
Each for $119.00
Add to Cart
 
11-101-8374
Accessories View Documents
MilliporeSigma Supelco
87472250MLF
250 mL
Each for $453.00
Add to Cart
 
Description

Description

Acylation, an alternative to silylation, is the conversion of compounds that contain active hydrogens (-OH, -SH and -NH) into esters, thioesters, and amides through the action of a carboxylic acid or derivative. The presence of a carbonyl group adjacent to the halogenated carbons enhances electron capture detector (ECD) response. Acylation has many benefits:. It improves the stability of compounds by protecting unstable groups.. It may confer volatility on substances such as carbohydrates or amino acids, which have so many polar groups that they are non-volatile and normally decompose on heating.. It assists in separations not possible with underivatized compounds.. Compounds are detectable at very low levels with an ECD.Hydrogen chloride-1-butanol is a reagent utilized to form fragmentation-directing derivatives for GC/MS analysis.
Specifications

Specifications

MFCD00011324
0.87 g/mL (at 20°C)
GC Derivatization
Videos
SDS
Documents

Documents

Product Certifications

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.