Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Hydrogen Chloride - 1-butanol Solution, ∼3 M in 1-butanol, For GC Derivatization, MilliporeSigma™ Supelco™

Catalog No. 111019704 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9704 50 mL
11-101-8374 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9704 Supplier MilliporeSigma Supelco Supplier No. 8747250MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acylation, an alternative to silylation, is the conversion of compounds that contain active hydrogens (-OH, -SH and -NH) into esters, thioesters, and amides through the action of a carboxylic acid or derivative. The presence of a carbonyl group adjacent to the halogenated carbons enhances electron capture detector (ECD) response. Acylation has many benefits:. It improves the stability of compounds by protecting unstable groups.. It may confer volatility on substances such as carbohydrates or amino acids, which have so many polar groups that they are non-volatile and normally decompose on heating.. It assists in separations not possible with underivatized compounds.. Compounds are detectable at very low levels with an ECD.Hydrogen chloride-1-butanol is a reagent utilized to form fragmentation-directing derivatives for GC/MS analysis.
TRUSTED_SUSTAINABILITY

Specifications

MDL Number MFCD00011324
Density 0.87 g/mL (at 20°C)
Grade GC Derivatization
Quantity 50 mL

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.