Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Methanesulfonic Acid, For HPLC, 99.5% (T), MilliporeSigma™ Supelco™

Catalog No. 111018808 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
10 x 1 mL
Packaging:
Clear Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-101-8808 10 x 1 mL Clear Glass Bottle
11-102-0290 1 mL Clear Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8808 Supplier MilliporeSigma Supelco Supplier No. 5951010X1ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75-75-2
Molecular Formula CH4O3S
Molecular Weight (g/mol) 96.10
MDL Number MFCD00007518
InChI Key AFVFQIVMOAPDHO-UHFFFAOYSA-N
IUPAC Name methanesulfonic acid
SMILES CS(O)(=O)=O

Specifications

Boiling Point 167°C (10 mmHg)
Melting Point 17°C to 19°C
Color APHA: ≤25
Quantity 10 x 1 mL
Linear Formula CH3SO3H
Formula Weight 96.11
Percent Purity ≥99.5% (T)
Grade HPLC
Packaging Clear Glass Bottle
Refractive Index n20/D 1.428-1.430; n20/D 1.429
Vapor Pressure 1 mmHg (20°C)
Density 1.481 g/mL (at 25°C); Vapor Density: 3.3 (vs. air)
Show More Show Less
RTECSNumber PB1140000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.