Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ N-Methyl-bis(trifluoroacetamide), 99.0%, MilliporeSigma™ Supelco™

Catalog No. 111018577 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8577 5 mL
11-101-7993 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8577 Supplier MilliporeSigma Supelco Supplier No. 659435ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 685-27-8
Product Line LiChropur
Density 1.547 g/mL (at 25°C)
Flash Point 45°C
Linear Formula CF3 CON(CH3)COCF3
MDL Number MFCD00000412
Boiling Point 121°C to 122°C
Synonym MBTFA
Type Organic Standard
Quantity 5 mL
Formula Weight 223.07
Percent Purity ≥99% (GC)
Grade GC/MS (SIM)
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.