Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 98.5%, MilliporeSigma™ Supelco™

Catalog No. 111019198 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9198 5 mL
11-101-8706 10 x 1 mL
11-101-8278 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-9198 Supplier MilliporeSigma Supelco Supplier No. 694795ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Methyl-N-(trimethylsilyl)trifluoroacetamide (MSTFA) is a silylating reagent.

TRUSTED_SUSTAINABILITY

Specifications

CAS 24589-78-4
Product Line LiChropur
Vapor Pressure 8.8 mmHg (27°C)
Vapor Density >1 (vs. air)
Density 1.075 g/mL (at 25°C)
Flash Point 26°C - closed cup
Linear Formula CF3 CON(CH3)Si(CH3)3
MDL Number MFCD00000411
Boiling Point 130°C to 132°C
Synonym N-Trimethylsilyl-N-methyl trifluoroacetamide; MSTFA
Type Organic Standard
Quantity 5 mL
Formula Weight 199.25
Percent Purity ≥98.5%
Grade GC Derivatization
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.