Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ N-Methyl-N-(trimethylsilyl)trifluoroacetamide, BioReagent for Silylations, MilliporeSigma™ Supelco™

Catalog No. 111019703 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9703 5 g
11-101-8422 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9703 Supplier MilliporeSigma Supelco Supplier No. M78915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00000411
Synonym N-Trimethylsilyl-N-methyl trifluoroacetamide; MSTFA

Specifications

CAS 24589-78-4
Density 1.07 g/mL at 25°C; 1.075 g/mL (at 25°C)
Boiling Point 130°C to 132°C
Flash Point 26°C
Packaging Ampule
Linear Formula CF3 CON(CH3)Si(CH3)3
Quantity 5 g
Molecular Weight (g/mol) 199.25
Vapor Density >1 (vs. air)
Formula Weight 199.25
Vapor Pressure 8.8 mmHg (27°C)
Grade Not Available
Physical Form Liquid
Product Line LiChropur
Chemical Name or Material N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Show More Show Less
RTECSNumber AC9800000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.