Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ N-Methyl-N-trimethylsilyltrifluoroacetamide Activated I, For GC Derivatization, MilliporeSigma™ Supelco™

Catalog No. 111017296 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7296 25 mL
11-101-7229 5 mL
11-101-7228 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-7296 Supplier MilliporeSigma Supelco Supplier No. 5099225MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Activated with Ethanethiol and Ammonium iodide

Derivatizing agent for the trimethylsilylation of -OH and -NH groups for GC and GC-MS
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Packaging Clear Glass Bottle
Refractive Index n20/D 1.380
Quantity 25 mL
Synonym MSTFA activated I
Grade For GC derivatization
Physical Form Solid

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.