Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, MilliporeSigma™ Supelco™

Catalog No. 111019764 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 x 1 mL
25 g
100 g
10 x 1 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9764 1 x 1 mL
11-101-9960 5 g
11-101-8754 10 x 1 mL
11-101-8761 25 g
11-101-8117 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 11-101-9764 Supplier MilliporeSigma Supelco Supplier No. T63811AMP
Add to Cart
Add to Cart

N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane is a silylating reagent generally used in estrogenic steroid hormone identification and quantification.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00008269
Synonym BSTFA + TMCS

Specifications

CAS 25561-30-2
Density 0.97 g/mL
Boiling Point 45°C to 50°C (14 mmHg)
Flash Point 6°C
Packaging Ampule
Linear Formula CF3 C[=NSi(CH3)3 ]OSi(CH3)3
Quantity 1 x 1 mL
Formula Weight 257.40
Grade For GC derivatization
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.