Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with tert-Butyldimethylchlorosilane, ≥95.0%, MilliporeSigma™

Catalog No. 111018247 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
10 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8247 10 mL
11-101-8159 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8247 Supplier MilliporeSigma Supelco Supplier No. 0094210ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00009671
Synonym N-Methyl-N-tert-butyldimethylsilyltrifluoroacetamide; MTBSTFA + 1% TBDMSCl; Silylating mixture VI

Specifications

CAS 77377-52-7
Flash Point 45°C - Closed Cup
Linear Formula C9H18F3 NOSi
Quantity 10 mL
Molecular Weight (g/mol) 241.33
Formula Weight 241.33
Percent Purity ≥95.0% (GC)
Grade For GC derivatization
Physical Form Liquid
Product Line LiChropur
Chemical Name or Material N-Methyl-N-tert-butyldimethylsilyltrifluoroacetamide
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.