Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ (R)-(+)-α-Methylbenzylamine, For chiral derivatization, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111018173 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8173 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8173 Supplier MilliporeSigma Supelco Supplier No. 7787925ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-(+)-alpha-Methylbenzylamine is a high quality, useful chiral derivatization reagent for all analytical applications, specific to GC in the chiral field. It is specially selected to meet the requirements for derivatization reagents for enantiomeric excess determinations.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00064405
Synonym (R)-(+)-1-Phenylethylamine

Specifications

CAS 3886-69-9
Density 0.952 g/mL (at 20°C)
Boiling Point 187°C to 189°C
Flash Point 70°C - Closed Cup
Linear Formula C6H5 CH(CH3)NH2
Quantity 25 mL
Formula Weight 121.18
Vapor Pressure 0.5 mmHg (20°C)
Percent Purity ≥99.0% (sum of enantiomers, GC); ≥99.0%
Grade For chiral derivatization
Product Line LiChropur
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.