Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Silylating Mixture III, For GC Derivatization, MilliporeSigma™ Supelco™

Catalog No. 111017217 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7217 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7217 Supplier MilliporeSigma Supelco Supplier No. 8543310ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Silylating mixture III is one of the most potent general silylating agents used for gas chromatography derivatization, comprising of N,O-bis(trimethylsilyl)trifluoroacetamide (BSTFA), 1-(trimethylsilyl)imidazole (TMSIM) and trimethylchlorosilane (TMCS) in the ratio 3:3:2.

Powerful and general silylating mixture
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mL
Synonym N,O-Bis-(trimethylsilyl)-trifluoracetamid - chlorotrimethylsilane - 1-(trimethylsilyl)imidazole mixture; 1-(Trimethylsilyl)imidazole/BSTFA/TMCS 3/3/2 (v/v/v); N,O-Bis(trimethylsilyl)trifluoroacetamide/TMSIM/TMCS 3/3/2 (v/v/v)
Grade GC Derivatization
Density 0.944 g/mL (at 20°C)

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.